2-Anisoyl chloride ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: 2-Methoxybenzoyl chloride, 2-Methoxybenzene-1-carbonyl chloride
M0108-25G M0108-500G M0108-100G
TCIAM0108-25GEA 910 SEK
TCIAM0108-25G TCIAM0108-500G TCIAM0108-100G
2-Anisoyl chloride ≥98.0% (by GC, titration analysis)
2-Anisoyl chloride
Formel: C₈H₇ClO₂
Kokpunkt: 140°C
Densitet: 1,57 g/cm³ (20 °C)
Flampunkt: 84°C
MDL Number: MFCD00000664
CAS nummer: 21615-34-9
EINECS: 244-477-5
UN: 1729
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eec14c496ccfa933e77989d0f80dc4de6acb6e9&vid=6eec14c496ccfa933e77989d0f80dc4de6acb6e9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %