4-Acetamidobenzoic acid ≥98.0%
Leverantör: TCI
|
Varning
|
A0013-25G
A0013-500G
TCIAA0013-25GEA
215
SEK
TCIAA0013-25G
TCIAA0013-500G
4-Acetamidobenzoic acid ≥98.0%
4-Acetamidobenzoic acid
Formel:
C₉H₉NO₃ Kokpunkt: 399 °C (1013 hPa) Smältpunkt: 259…262 °C |
MDL Number:
MFCD00002534 CAS nummer: 556-08-1 EINECS: 209-114-7 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...