4-Chlorobenzenesulphonyl chloride ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 4-Chlorobenzene-1-sulphonyl chloride
C0128-25G C0128-500G
TCIAC0128-25GEA 255 SEK
TCIAC0128-25G TCIAC0128-500G
4-Chlorobenzenesulphonyl chloride ≥98.0%
4-Chlorobenzenesulphonyl chloride
Formel: C₆H₄Cl₂O₂S
MW: 211,07 g/mol
Kokpunkt: 141°C
Smältpunkt: 51 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007439
CAS nummer: 98-60-2
EINECS: 202-685-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56a18bece7f831af171b02d0878a386f12f03061&vid=56a18bece7f831af171b02d0878a386f12f03061&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 51.0 to 56.0 deg-C
Solubility in Toluene almost transparency