2,4,5-Trichlorobenzenesulphonyl chloride ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2,4,5-Trichlorobenzene-1-sulphonyl chloride

B0247-25G B0247-500G
TCIAB0247-25GEA 685 SEK
TCIAB0247-25G TCIAB0247-500G
2,4,5-Trichlorobenzenesulphonyl chloride ≥98.0%
2,4,5-Trichlorobenzenesulphonyl chloride
Formel: C₆H₂Cl₄O₂S
MW: 279,96 g/mol
Smältpunkt: 69 °C
Densitet: 1,88 g/cm³ (20 °C)
MDL Number: MFCD00007428
CAS nummer: 15945-07-0
EINECS: 240-079-0
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2b3fb0cc95cf9b5972f55f712aa31aa41a7ad4a&vid=b2b3fb0cc95cf9b5972f55f712aa31aa41a7ad4a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Toluene very faint turbidity
Melting point 65.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR