1-Chloro-3,5-dimethoxybenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Chloro-1,3-dimethoxybenzene
C1258-10G
C1258-25G
TCIAC1258-10GEA
1300
SEK
TCIAC1258-10G
TCIAC1258-25G
1-Chloro-3,5-dimethoxybenzene ≥97.0%
1-Chloro-3,5-dimethoxybenzene
Formel:
C₈H₉ClO₂ MW: 172,61 g/mol Kokpunkt: 113°C Smältpunkt: 38 °C |
MDL Number:
MFCD00008382 CAS nummer: 7051-16-3 EINECS: 230-330-2 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...