1-Chloro-2,5-dimethoxybenzene ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 2-Chloro-1,4-dimethoxybenzene

C1577-25G C1577-500G
TCIAC1577-25GEA 215 SEK
TCIAC1577-25G TCIAC1577-500G
1-Chloro-2,5-dimethoxybenzene ≥97.0%
1-Chloro-2,5-dimethoxybenzene
Formel: C₈H₉ClO₂
MW: 172,61 g/mol
Kokpunkt: 235°C
Smältpunkt: 8,1 °C
Densitet: 1,21 g/cm³ (20 °C)
Flampunkt: 123°C
MDL Number: MFCD00008356
CAS nummer: 2100-42-7
EINECS: 218-267-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4793e7952f0ed857061dc6007dbf5967f52dc987&vid=4793e7952f0ed857061dc6007dbf5967f52dc987&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR