(2-Chloroethoxy)benzene ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: ω-Chlorophenetole, 2-Chloroethyl phenyl ether, β-Chlorophenetole, 2-Phenoxyethyl chloride

C0240-25ML C0240-500ML
TCIAC0240-25MLEA 510 SEK
TCIAC0240-25ML TCIAC0240-500ML
(2-Chloroethoxy)benzene ≥95.0%
(2-Chloroethoxy)benzene
Formel: C₆H₅OCH₂CH₂Cl
MW: 156,61 g/mol
MDL Number: MFCD00018946
CAS nummer: 622-86-6
EINECS: 210-757-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be79065e42e801268c3bf9e4b9ae8eece6a9b6a2&vid=be79065e42e801268c3bf9e4b9ae8eece6a9b6a2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR