4-Bromobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
B0550-25G B0550-500G
TCIAB0550-25GEA 935 SEK
TCIAB0550-25G TCIAB0550-500G
4-Bromobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
4-Bromobenzenesulphonyl chloride
Formel: C₆H₄BrClO₂S
MW: 255,52 g/mol
Kokpunkt: 153°C
Smältpunkt: 74 °C
Densitet: 1,37 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007437
CAS nummer: 98-58-8
EINECS: 202-683-2
UN: 3261
ADR: 8,III
Merck Index: 14,01407
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e2c897fd6aa7ed07b02520a854e8d751bc419d&vid=f9e2c897fd6aa7ed07b02520a854e8d751bc419d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 74.0 to 77.0 deg-C
Solubility in Toluene almost transparency