4-Hydroxy-1-methylpiperidine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
M0933-25G M0933-500G
TCIAM0933-25GEA 215 SEK
TCIAM0933-25G TCIAM0933-500G
4-Hydroxy-1-methylpiperidine ≥98.0% (by GC, titration analysis)
4-Hydroxy-1-methylpiperidine
Formel: C₆H₁₃NO
Kokpunkt: 105°C
Smältpunkt: 29…31 °C
Densitet: 0,98 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006500
CAS nummer: 106-52-5
EINECS: 203-406-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ccaeefb8c9ec5f15c4a756debc13609aee5bf45&vid=2ccaeefb8c9ec5f15c4a756debc13609aee5bf45&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %