6-Phenylcaproic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Phenylhexanoic acid
P1108-25G
P1108-5G
TCIAP1108-25GEA
6750
SEK
TCIAP1108-25G
TCIAP1108-5G
6-Phenylcaproic acid ≥98.0% (by GC, titration analysis)
6-Phenylcaproic acid
Formel:
C₁₂H₁₆O₂ MW: 192,26 g/mol Kokpunkt: 201 - 202 °C (24 -rr) Smältpunkt: 23 °C Densitet: 1,02 g/cm³ (20 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00014381 CAS nummer: 5581-75-9 |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...