Methyl pivalate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Methyl trimethylacetate
P0462-25ML P0462-500ML
TCIAP0462-25MLEA 215 SEK
TCIAP0462-25ML TCIAP0462-500ML
Methyl pivalate ≥98.0%
Methyl pivalate
Formel: C₆H₁₂O₂
MW: 116,16 g/mol
Kokpunkt: 102°C
Smältpunkt: < –70 °C
Densitet: 0,87 g/cm³ (20 °C)
Flampunkt: 10°C
Storage Temperature: Omgivande
MDL Number: MFCD00008843
CAS nummer: 598-98-1
EINECS: 209-959-1
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9472baf0c33b86c3753cdbb02e29baa2ed5cd31&vid=b9472baf0c33b86c3753cdbb02e29baa2ed5cd31&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %