Methyl methanesulphonate ≥98.0%

Leverantör: TCI
Fara

M0369-25G M0369-100G
TCIAM0369-25GEA 765 SEK
TCIAM0369-25G TCIAM0369-100G
Methyl methanesulphonate ≥98.0%
Methyl methanesulphonate
Formel: C₂H₆O₃S
Kokpunkt: 203 °C (1013 hPa)
Smältpunkt: 20 °C
Densitet: 1,3 g/cm³ (20 °C)
Flampunkt: 104°C
Storage Temperature: Omgivande
MDL Number: MFCD00007557
CAS nummer: 66-27-3
EINECS: 200-625-0
UN: 2810
ADR: 6.1,II
Merck Index: 14,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e29f9f38f36ac29ba527deff26c1c2777a8c371&vid=7e29f9f38f36ac29ba527deff26c1c2777a8c371&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR