2-Nitroethanol ≥93.0%
Leverantör: TCI EUROPE
|
Varning
|
N0368-25G
N0368-5G
TCIAN0368-25GEA
2950
SEK
TCIAN0368-25G
TCIAN0368-5G
2-Nitroethanol ≥93.0%
2-Nitroethanol
Formel:
C₂H₅NO₃ MW: 91,07 g/mol Kokpunkt: 197°C Smältpunkt: –80 °C Densitet: 1,29 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00007405 CAS nummer: 625-48-9 EINECS: 210-895-1 |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...