4-(Dimethylamino)-1-butanol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 4-(Dimethylamino)butan-1-ol, 4-Dimethylamino-1-butanol
D1659-25ML D1659-5ML
TCIAD1659-25MLEA 2025 SEK
TCIAD1659-25ML TCIAD1659-5ML
4-(Dimethylamino)-1-butanol ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)-1-butanol
Formel: C₆H₁₅NO
MW: 117,19 g/mol
Kokpunkt: 188°C
Densitet: 0,88 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00020624
CAS nummer: 13330-96-6
EINECS: 236-380-1
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65629f511cf4e56912dd93266d258fadc53bdf19&vid=65629f511cf4e56912dd93266d258fadc53bdf19&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %