1-Amino-2,2,2-trifluoroethane ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 2,2,2-Trifluoroethylamine
T1169-25G T1169-5G
TCIAT1169-25GEA 670 SEK
TCIAT1169-25G TCIAT1169-5G
1-Amino-2,2,2-trifluoroethane ≥97.0% (by GC, titration analysis)
1-Amino-2,2,2-trifluoroethane
Formel: C₂H₄F₃N
MW: 99,06 g/mol
Kokpunkt: 37°C
Densitet: 1,2452 g/cm³ (25 °C)
Flampunkt: -17°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008132
CAS nummer: 753-90-2
EINECS: 212-041-3
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0b9308d9836453c02f0cd7b31d9b540ea1fd4ce&vid=b0b9308d9836453c02f0cd7b31d9b540ea1fd4ce&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %