Thioacetic acid ≥95.0%

Leverantör: TCI EUROPE
Fara

T0189-25ML T0189-500ML T0189-100ML
TCIAT0189-25MLEA 215 SEK
TCIAT0189-25ML TCIAT0189-500ML TCIAT0189-100ML
Thioacetic acid ≥95.0%
Thioacetic acid
Formel: C₂H₄OS
MW: 76,12 g/mol
Kokpunkt: 93°C
Smältpunkt: –17 °C
Densitet: 1,07 g/cm³ (20 °C)
Flampunkt: 5°C
Storage Temperature: Kylskåp
MDL Number: MFCD00004853
CAS nummer: 507-09-5
EINECS: 208-063-8
UN: 2436
ADR: 3,II
Merck Index: 14,09320
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32297d6cc0c026f915c9853b78490b2dd433c038&vid=32297d6cc0c026f915c9853b78490b2dd433c038&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR