Butyl laurate ≥99.0%
Leverantör: TCI EUROPE
Synonyms: Butyl dodecanoate, Dodecylic acid butyl ester, Dodecanoic acid butyl ester, Lauric acid butyl ester, n-Butyl dodecanoate
L0083-25ML L0083-500ML
TCIAL0083-25MLEA 230 SEK
TCIAL0083-25ML TCIAL0083-500ML
Butyl laurate ≥99.0%
Butyl laurate
Formel: C₁₆H₃₂O₂
MW: 256,43 g/mol
Kokpunkt: 199°C
Flampunkt: 166°C
MDL Number: MFCD00042870
CAS nummer: 106-18-3
EINECS: 203-370-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a96a1f09c0c78a2bae1137aafd2daabfd61a187a&vid=a96a1f09c0c78a2bae1137aafd2daabfd61a187a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %