2-Chloroethyl isothiocyanate ≥98.0%

Leverantör: TCI EUROPE
Fara

C2245-1G C2245-5G
TCIAC2245-1GEA 230 SEK
TCIAC2245-1G TCIAC2245-5G
2-Chloroethyl isothiocyanate ≥98.0%
2-Chloroethyl isothiocyanate
Formel: C₃H₄ClNS
MW: 121,59 g/mol
Kokpunkt: 60°C
Densitet: 1,265 g/cm³ (20 °C)
Flampunkt: 43°C
MDL Number: MFCD00041125
CAS nummer: 6099-88-3
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=679b28d467b5aff49c653f5ffe3e998878a62c7c&vid=679b28d467b5aff49c653f5ffe3e998878a62c7c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR