4'-Chloro-2'-fluoroacetanilide ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: N-(4-Chloro-2-fluorophenyl)acetamide
C1692-25G C1692-5G
TCIAC1692-25GEA 335 SEK
TCIAC1692-25G TCIAC1692-5G
4'-Chloro-2'-fluoroacetanilide ≥98.0%
4'-Chloro-2'-fluoroacetanilide
Formel: C₈H₇ClFNO
MW: 187,6 g/mol
Kokpunkt: 332 °C (1013 hPa)
Smältpunkt: 156…158 °C
MDL Number: MFCD00042520
CAS nummer: 59280-70-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a5e9fb59860dab9a67b93f19b99e684c94afd4e&vid=7a5e9fb59860dab9a67b93f19b99e684c94afd4e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 155.0 to 159.0 deg-C