2'-Chloro-4'-fluoroacetanilide ≥98.0%

Leverantör: TCI
Varning

Synonyms: N-(2-Chloro-4-fluorophenyl)acetamide

C1311-25G C1311-5G
TCIAC1311-25GEA 1625 SEK
TCIAC1311-25G TCIAC1311-5G
2'-Chloro-4'-fluoroacetanilide ≥98.0%
2'-Chloro-4'-fluoroacetanilide
Formel: C₈H₇ClFNO
Kokpunkt: 332 °C (1013 hPa)
Smältpunkt: 117 °C
MDL Number: MFCD00042594
CAS nummer: 399-35-9

Order Now

Specifikation testresultat

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR