Ethyl bromofluoroacetate ≥98.0%
Leverantör: TCI
Fara
Synonyms: Bromofluoroacetic ethyl ester, Ethylbromofluoroacetate
B1673-25G B1673-5G
TCIAB1673-25GEA 1700 SEK
TCIAB1673-25G TCIAB1673-5G
Ethyl bromofluoroacetate ≥98.0%
Ethyl bromofluoroacetate
Formel: BrFCHCOOCH₂CH₃
Kokpunkt: 68°C
Densitet: 1,55866 g/cm³ (17 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00042095
CAS nummer: 401-55-8
EINECS: 206-928-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34958b6d39d49db50b92b45a57ca22865ccf3022&vid=34958b6d39d49db50b92b45a57ca22865ccf3022&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %