6-Bromocaproic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
6-Bromohexanoic acid
B1290-25G
B1290-250G
TCIAB1290-25GEA
525
SEK
TCIAB1290-25G
TCIAB1290-250G
6-Bromocaproic acid ≥98.0% (by GC, titration analysis)
6-Bromocaproic acid
Formel:
BrH₂C(CH₂)₄COOH MW: 195,06 g/mol Kokpunkt: 170°C Smältpunkt: 31…34 °C Densitet: 1,4068 g/cm³ (30 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00004422 CAS nummer: 4224-70-8 EINECS: 224-176-5 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 34.0 to 37.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...