4'-Bromoacetanilide ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Bromoacetanilide, N-(4-Bromophenyl)acetamide
B0534-25G
B0534-500G
B0534-100G
TCIAB0534-25GEA
405
SEK
TCIAB0534-25G
TCIAB0534-500G
TCIAB0534-100G
4'-Bromoacetanilide ≥98.0%
4'-Bromoacetanilide
Formel:
H₃CC(O)NH(C₆H₄Br) MW: 214,06 g/mol Kokpunkt: 347 °C (1013 hPa) Smältpunkt: 164…167 °C Densitet: 1,717 g/cm³ (20 °C) |
MDL Number:
MFCD00000092 CAS nummer: 103-88-8 EINECS: 203-154-9 Merck Index: 14,01399 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 166.0 to 172.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...