Bromopyruvic acid ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid
B1153-25G
B1153-5G
TCIAB1153-25GEA
3900
SEK
TCIAB1153-25G
TCIAB1153-5G
Bromopyruvic acid ≥98.0%
Bromopyruvic acid
Formel:
BrH₂CC(O)COOH MW: 166,96 g/mol Kokpunkt: 249 °C (1013 hPa) Smältpunkt: 77…79 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00002587 CAS nummer: 1113-59-3 EINECS: 214-206-5 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 98.0 to 103.0 % |
Melting point | 74.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...