5-Chloro-1-phenyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
5-Chloro-1-phenyltetrazole
C0999-25G
C0999-5G
TCIAC0999-25GEA
1475
SEK
TCIAC0999-25G
TCIAC0999-5G
5-Chloro-1-phenyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-Chloro-1-phenyl-1H-tetrazole
Formel:
C₇H₅ClN₄ MW: 180,6 g/mol Smältpunkt: 121…123 °C |
MDL Number:
MFCD00003128 CAS nummer: 14210-25-4 EINECS: 238-065-4 UN: 1325 ADR: 4.1,III |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...