4,7-Difenyl-1,10-fenantrolin ≥99.0%

Leverantör: TCI EUROPE

Synonyms: 4,7-Diphenyl-1,10-phenanthroline, (Bathophenanthroline)

D0905-1G D0905-5G
TCIAD0905-1GEA 1425 SEK
TCIAD0905-1G TCIAD0905-5G
4,7-Difenyl-1,10-fenantrolin ≥99.0%
4,7-Difenyl-1,10-fenantrolin
Formel: C₂₄H₁₆N₂
MW: 332,4 g/mol
Kokpunkt: 508 °C (1013 hPa)
Smältpunkt: 215…220 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004976
CAS nummer: 1662-01-7
EINECS: 216-767-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77dde1e3336d48b30fd99e5b71e11ae508384335&vid=77dde1e3336d48b30fd99e5b71e11ae508384335&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 218.0 to 221.0 deg-C
Solubility in hot EtOH almost transparency
Sensitiveness Abs min.0.36 (near 533nm) in the presence of Fe (1 ppm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR