L (+)-Norvalin 99,0 Titrimetrisk
Leverantör: TCI
Synonyms:
(S)-(+)-Norvaline, L(+)-Norvaline, H-Nva-OH, (S)-2-Aminopentanoic acid
N0686-1G
N0686-5G
TCIAN0686-1GEA
245
SEK
TCIAN0686-1G
TCIAN0686-5G
L (+)-Norvalin 99,0 Titrimetrisk
L (+)-Norvalin
Formel:
C₅H₁₁NO₂ MW: 117.15 g/mol Kokpunkt: 228 Smältpunkt: 101 |
MDL Number:
MFCD00064421 CAS nummer: 6600-40-4 EINECS: 229-543-3 Merck Index: 14,06716 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | +24.0 to +26.0 deg (C=10, 6mol/L HCl) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...