(S)-(+)-2-Amino-1-butanol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol
A0973-1ML A0973-5ML
TCIAA0973-1MLEA 230 SEK
TCIAA0973-1ML TCIAA0973-5ML
(S)-(+)-2-Amino-1-butanol ≥98.0% (by GC, titration analysis)
(S)-(+)-2-Amino-1-butanol
Formel: C₄H₁₁NO
MW: 89,14 g/mol
Kokpunkt: 178°C
Smältpunkt: –2 °C
Densitet: 0,947 g/cm³ (20 °C)
Flampunkt: 79°C
Storage Temperature: Omgivande
MDL Number: MFCD00064418
CAS nummer: 5856-62-2
EINECS: 227-475-9
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec8d9dea66693f185fd2eb51717b782058f8793b&vid=ec8d9dea66693f185fd2eb51717b782058f8793b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +9.0 to +11.0 deg (neat)