(±)-1-Heptyn-3-ol ≥97.0%

Leverantör: TCI

H0455-1ML H0455-5ML
TCIAH0455-1MLEA 215 SEK
TCIAH0455-1ML TCIAH0455-5ML
(±)-1-Heptyn-3-ol ≥97.0%
(±)-1-Heptyn-3-ol
Formel: C₇H₁₂O
Kokpunkt: 73°C
Densitet: 0,87 g/cm³ (20 °C)
Flampunkt: 65°C
Storage Temperature: Kylskåp
MDL Number: MFCD00041597
CAS nummer: 7383-19-9
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e45b1dca863b07b84c5401fc06f4f0a2e419af8b&vid=e45b1dca863b07b84c5401fc06f4f0a2e419af8b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR