(±)-1-Octen-3-ol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (±)-Oct-1-en-3-ol, (±)-Pentyl vinyl carbinol, Oct-1-en-3-ol

O0159-25ML O0159-100ML
TCIAO0159-25MLEA 415 SEK
TCIAO0159-25ML TCIAO0159-100ML
(±)-1-Octen-3-ol ≥98.0%
(±)-1-Octen-3-ol
Formel: C₈H₁₆O
MW: 128,21 g/mol
Kokpunkt: 166°C
Smältpunkt: –49 °C
Densitet: 0,83 g/cm³ (20 °C)
Flampunkt: 63°C
Storage Temperature: Omgivande
MDL Number: MFCD00004589
CAS nummer: 3391-86-4
EINECS: 222-226-0
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf21efc73d03cd1c823e1fd8ab401b19c8a7363c&vid=bf21efc73d03cd1c823e1fd8ab401b19c8a7363c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR