(±)-2-Phenylbutyric acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (±)-2-Phenylbutanoic acid, 2-Phenylbutanoic acid

P0164-25G P0164-500G
TCIAP0164-25GEA 350 SEK
TCIAP0164-25G TCIAP0164-500G
(±)-2-Phenylbutyric acid ≥98.0% (by GC, titration analysis)
(±)-2-Phenylbutyric acid
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Kokpunkt: 272°C
Smältpunkt: 39…42 °C
MDL Number: MFCD00002667
CAS nummer: 90-27-7
EINECS: 201-982-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83cd335305fefe69a723157abddfd4c1c62edf56&vid=83cd335305fefe69a723157abddfd4c1c62edf56&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 42.0 to 45.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR