DL-Valin ≥98.0%

Leverantör: TCI EUROPE

Synonyms: H-DL-Val-OH, (±)-Valine, 2-Amino-3-methylbutanoic acid

V0013-25G V0013-500G
TCIAV0013-25GEA 230 SEK
TCIAV0013-25G TCIAV0013-500G
DL-Valin ≥98.0%
DL-Valin
Formel: C₅H₁₁NO₂
MW: 117,15 g/mol
Kokpunkt: 217 °C (1013 hPa)
Smältpunkt: 86 °C
Densitet: 1,352 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004267
CAS nummer: 516-06-3
EINECS: 208-220-0
Merck Index: 14,09909
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ae04340ba30342b4273cc928645a0883412b79a&vid=0ae04340ba30342b4273cc928645a0883412b79a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR