(±)-2-Tridecanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
2-Tridecanol, (±)-2-Hydroxy tridecane
T0633-25G
T0633-5G
TCIAT0633-25GEA
15225
SEK
TCIAT0633-25G
TCIAT0633-5G
(±)-2-Tridecanol ≥98.0%
(±)-2-Tridecanol
Formel:
C₁₃H₂₈O MW: 200,36 g/mol Kokpunkt: 265°C Smältpunkt: 23 °C Densitet: 0,8215 g/cm³ (34 °C) |
MDL Number:
MFCD00037601 CAS nummer: 1653-31-2 EINECS: 216-723-1 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...