(±)-Mercaptosuccinic acid ≥98.0%

Leverantör: TCI
Varning

Synonyms: (±)-Mercapto-1,4-butanedioic acid, 2-Mercaptosuccinic acid, (±)-Thiomalic acid, DL-Mercaptosuccinic acid

M0064-25G M0064-500G M0064-100G
TCIAM0064-25GEA 270 SEK
TCIAM0064-25G TCIAM0064-500G TCIAM0064-100G
(±)-Mercaptosuccinic acid ≥98.0%
(±)-Mercaptosuccinic acid
Formel: C₄H₆O₄S
MW: 150.16 g/mol
Kokpunkt: 322
Storage Temperature: Omgivande
MDL Number: MFCD00004860
CAS nummer: 70-49-5
EINECS: 200-736-4
Merck Index: 14,09343
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=672051bba62b0f9fbb28e3f8ccff6f2d2a02caa9&vid=672051bba62b0f9fbb28e3f8ccff6f2d2a02caa9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 152.0 to 155.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR