DL-Homocysteine ≥90.0%

Leverantör: TCI

Synonyms: (±)-Homocysteine, 2-Amino-4-mercaptobutanoic acid, H-DL-HoCys-OH

H0159-1G H0159-5G
TCIAH0159-1GEA 910 SEK
TCIAH0159-1G TCIAH0159-5G
DL-Homocysteine ≥90.0%
DL-Homocysteine
Formel: C₄H₉NO₂S
Storage Temperature: Frys
MDL Number: MFCD00004898
CAS nummer: 454-29-5
EINECS: 207-222-9
Merck Index: 14,04734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=135b293a1a464559cb498af13eb5844d452982a2&vid=135b293a1a464559cb498af13eb5844d452982a2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR