Fullerene C60 ≥99.0%
Leverantör: TCI EUROPE
Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen
B1660-1G B1660-100MG
TCIAB1660-1GEA 1825 SEK
TCIAB1660-1G TCIAB1660-100MG
Fullerene C60 ≥99.0%
Fullerene C60
Formel: C₆₀
MW: 720,66 g/mol
Smältpunkt: > 280 °C
Densitet: ~ 1,65 g/cm³ (20 °C)
MDL Number: MFCD00151408
CAS nummer: 99685-96-8
ADR: 4.1,III
Merck Index: 14,01462
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9621e9e818dd79e0860d45aa7fa0184e7954f5d&vid=f9621e9e818dd79e0860d45aa7fa0184e7954f5d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Gray to Dark blue to Black powder to crystaline
Purity (HPLC) min. 99.0 area%
Absorbance (E1%1cm) min. 750 (335.5nm)