Triisopropylsilane ≥98.0%
Leverantör: TCI
|
Varning
|
T1533-100ML
T1533-25ML
T1533-5ML
TCIAT1533-100MLEA
885
SEK
TCIAT1533-100ML
TCIAT1533-25ML
TCIAT1533-5ML
Triisopropylsilane ≥98.0%
Triisopropylsilane
Formel:
C₉H₂₂Si MW: 158.36 g/mol Kokpunkt: 166°C Flampunkt: 37°C |
MDL Number:
MFCD00009657 CAS nummer: 6485-79-6 EINECS: 464-880-1 UN: 1993 ADR: 3 III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...