Tetramethylgermane 98,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
T1158-1G
T1158-5G
TCIAT1158-1GEA
485
SEK
TCIAT1158-1G
TCIAT1158-5G
Tetramethylgermane 98,0 GC_ANALYSMETOD
Tetramethylgermane
Formel:
C₄H₁₂Ge MW: 132.75 g/mol Kokpunkt: 43°C Smältpunkt: -88 Densitet: 0.9781 Flampunkt: –37°C |
MDL Number:
MFCD00014843 CAS nummer: 865-52-1 EINECS: 212-745-0 UN: 1993 ADR: 3 II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...