Tetrafluorosuccinic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2,2,3,3-Tetrafluorosuccinic acid, Tetrafluoro-1,4-butanedioic acid

T1621-25G T1621-5G
TCIAT1621-25GEA 2775 SEK
TCIAT1621-25G TCIAT1621-5G
Tetrafluorosuccinic acid ≥98.0%
Tetrafluorosuccinic acid
Formel: C₄H₂F₄O₄
MW: 190,05 g/mol
Kokpunkt: 120°C
Smältpunkt: 118…120 °C
Densitet: 1,4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00042430
CAS nummer: 377-38-8
EINECS: 206-820-7
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32facae804993b37f2c7121b56f481ac1806e9b6&vid=32facae804993b37f2c7121b56f481ac1806e9b6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 121.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR