Tween® 60 (Polysorbate)
Leverantör: TCI EUROPE
Synonyms: Polyoxyethylene sorbitan monostearate
T0545-25G T0545-500G
TCIAT0545-25GEA 215 SEK
TCIAT0545-25G TCIAT0545-500G
Tween® 60 (Polysorbate)
Tween® 60 (Polysorbate)
Formel: C₆₄H₁₂₆O₂₆
Kokpunkt: ? 110 °C (1013 hPa)
Densitet: 1,08 g/cm³ (20 °C)
Flampunkt: ≥ 149 °C
Storage Temperature: Omgivande
MDL Number: MFCD01780110
CAS nummer: 9005-67-8
EINECS: 500-020-4
Order Now
Specifikation testresultat
Appearance White or Colorless to Yellow powder to lump to clear liquid
Freezing point 21.0 to 25.0 deg-C
Saponification Value 43.0 to 55.0