Propionylklorid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Propanoylklorid
P0516-25G P0516-500G
TCIAP0516-25GEA 215 SEK
TCIAP0516-25G TCIAP0516-500G
Propionylklorid ≥98.0% (by GC, titration analysis)
Propionylklorid
Formel: C₃H₅ClO
MW: 92,52 g/mol
Kokpunkt: 80°C
Smältpunkt: –94°C
Densitet: 1,06 g/cm³ (20 °C)
Flampunkt: 12°C
Storage Temperature: Omgivande
MDL Number: MFCD00000745
CAS nummer: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3,II
Merck Index: 14,07828
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=271e5aa0e0c4384ba8a7326dd4eb5ad8f35cc2d8&vid=271e5aa0e0c4384ba8a7326dd4eb5ad8f35cc2d8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %