Polyetylenglykol 300

Leverantör: TCI EUROPE

Synonyms: PEG 300

H0543-25G H0543-500G
TCIAH0543-25GEA 215 SEK
TCIAH0543-25G TCIAH0543-500G
Polyetylenglykol 300
Polyetylenglykol 300
Formel: H(OCH₂CH₂)nOH
MW: 300 g/mol
Smältpunkt: –15…–10 °C
Densitet: 1,13 g/cm³ (20 °C)
Flampunkt: 196°C
Storage Temperature: Omgivande
MDL Number: MFCD00081839
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,07568

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Average molecular weight 290 to 305

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR