Polyetylenglykol 200
Leverantör: TCI
Synonyms:
PEG 200, Ethylene glycol polymer average Mn 200
P0840-25G
P0840-500G
TCIAP0840-25GEA
215
SEK
TCIAP0840-25G
TCIAP0840-500G
Polyetylenglykol 200
Polyetylenglykol 200
Formel:
H(OCH₂CH₂)nOH MW: 200 g/mol Kokpunkt: ? 250 °C (1013 hPa) Smältpunkt: –55…–40 °C Densitet: 1,124 g/cm³ (20 °C) Flampunkt: 171°C Storage Temperature: Omgivande |
MDL Number:
MFCD01779596 CAS nummer: 25322-68-3 EINECS: 500-038-2 Merck Index: 14,07568 |
Specifikation testresultat
Appearance | Colorless clear liquid |
Average molecular weight | 190 to 210 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...