Bifenyl ≥99.5%
Leverantör: TCI EUROPE
Varning
Synonyms: Difenyl
B0465-25G B0465-500G
TCIAB0465-25GEA 215 SEK
TCIAB0465-25G TCIAB0465-500G
Bifenyl ≥99.5%
Bifenyl
Formel: C₁₂H₁₀
MW: 154,21 g/mol
Kokpunkt: 256°C
Smältpunkt: 68…70 °C
Densitet: 1,175 g/cm³ (20 °C)
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003054
CAS nummer: 92-52-4
EINECS: 202-163-5
UN: 3077
ADR: 9,III
Merck Index: 14,03314
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=417ad70b9bfe414891502b3b2a8630510216507f&vid=417ad70b9bfe414891502b3b2a8630510216507f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to lump
Purity (GC) min. 99.5 %
Melting point 69.0 to 72.0 deg-C
Solubility in EtOH almost transparency