Ethyl valerate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Ethyl Pentanoate, Valeric acid ethyl ester
V0004-25ML V0004-500ML
TCIAV0004-25MLEA 215 SEK
TCIAV0004-25ML TCIAV0004-500ML
Ethyl valerate ≥98.0%
Ethyl valerate
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 145°C
Smältpunkt: –91 °C
Densitet: 0,87 g/cm³ (20 °C)
Flampunkt: 41°C
Storage Temperature: Omgivande
MDL Number: MFCD00009479
CAS nummer: 539-82-2
EINECS: 208-726-1
UN: 3272
ADR: 3,III
Merck Index: 12,10042
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %