Decafluorobenzophenone ≥98.0%

Leverantör: TCI

D1631-1G D1631-5G
TCIAD1631-1GEA 685 SEK
TCIAD1631-1G TCIAD1631-5G
Decafluorobenzophenone ≥98.0%
Decafluorobenzophenone
Formel: C₁₃F₁₀O
Kokpunkt: 263 °C (1013 hPa)
Smältpunkt: 94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000295
CAS nummer: 853-39-4
EINECS: 212-717-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=883cf6030d3adf33abb6eaac783a25e3ebadac6b&vid=883cf6030d3adf33abb6eaac783a25e3ebadac6b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystaline
Purity (GC) min. 98.0 %
Melting point 91.0 to 95.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR