Trimetylfosfat ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Trimethylphosphate
P0271-25G
P0271-500G
TCIAP0271-25GEA
215
SEK
TCIAP0271-25G
TCIAP0271-500G
Trimetylfosfat ≥98.0%
Trimetylfosfat
Formel:
C₃H₉O₄P MW: 140,08 g/mol Kokpunkt: 197°C Smältpunkt: –46°C Densitet: 1,21 g/cm³ (20 °C) Flampunkt: 107°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008348 CAS nummer: 512-56-1 EINECS: 208-144-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...