Octyltriethoxysilane ≥97.0%

Leverantör: TCI
Varning

O0171-25ML O0171-500ML O0171-100ML
TCIAO0171-25MLEA 215 SEK
TCIAO0171-25ML TCIAO0171-500ML TCIAO0171-100ML
Octyltriethoxysilane ≥97.0%
Octyltriethoxysilane
Formel: C₁₄H₃₂O₃Si
Kokpunkt: 106°C
Smältpunkt: –40 °C
Densitet: 0,8515 g/cm³ (20 °C)
Flampunkt: 100°C
Storage Temperature: Omgivande
MDL Number: MFCD00039883
CAS nummer: 2943-75-1
EINECS: 220-941-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3d809a9193063cca5b476c0643e896febb13b7a&vid=f3d809a9193063cca5b476c0643e896febb13b7a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR