Methyltrimethoxysilane ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Trimethoxy(methyl)silane
M0660-25ML
M0660-500ML
M0660-100ML
TCIAM0660-25MLEA
215
SEK
TCIAM0660-25ML
TCIAM0660-500ML
TCIAM0660-100ML
Methyltrimethoxysilane ≥98.0%
Methyltrimethoxysilane
Formel:
C₄H₁₂O₃Si MW: 136,22 g/mol Kokpunkt: 103°C Smältpunkt: 11 °C Densitet: 0,955 g/cm³ (20 °C) Flampunkt: 13°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008342 CAS nummer: 1185-55-3 EINECS: 214-685-0 UN: 1993 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...