Metylformiat ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
Formic acid methyl ester
F0057-25ML
F0057-500ML
TCIAF0057-25MLEA
215
SEK
TCIAF0057-25ML
TCIAF0057-500ML
Metylformiat ≥95.0%
Metylformiat
Formel:
C₂H₄O₂ MW: 60.05 g/mol Kokpunkt: 32°C Smältpunkt: -100.4 Flampunkt: –20°C Storage Temperature: Omgivande |
MDL Number:
MFCD00003291 CAS nummer: 107-31-3 EINECS: 203-481-7 UN: 1243 ADR: 3 I Merck Index: 14,06077 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...